Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STARD5 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501985753
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-5753 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-5753 Supplier Bioss Supplier No. BS3919R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

May be involved in the intracellular transport of sterols or other lipids. May bind cholesterol or other sterols.

Specifications

Antigen STARD5
Applications ELISA, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene STARD5
Gene Accession No. Q9NSY2
Gene Alias 18B7-T7(GS); 2310058G22Rik; AI607880; D7Ertd152e; StAR related lipid transfer domain containing 5; Stard5; StAR-related lipid transfer (START) domain containing 5; StAR-related lipid transfer domain containing 5; StAR-related lipid transfer protein 5; StAR-related protein 1-4E; StAR-related protein p3-15E/p3-16E; START domain containing 5; START domain-containing protein 5
Gene Symbols STARD5
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human STARD5.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 80765
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.