Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ STARD6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA548291
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA548291 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA548291 Supplier Invitrogen™ Supplier No. PA548291
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

STARD6 Polyclonal Antibody for Western Blot, Flow

May be involved in the intracellular transport of sterols or other lipids. May bind cholesterol or other sterols.
TRUSTED_SUSTAINABILITY

Specifications

Antigen STARD6
Applications Flow Cytometry, Western Blot
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene STARD6
Gene Accession No. P59095
Gene Alias StAR related lipid transfer domain containing 6; STARD6; StAR-related lipid transfer (START) domain containing 6; StAR-related lipid transfer domain containing 6; stAR-related lipid transfer protein 6; START domain containing 6; START domain-containing protein 6
Gene Symbols STARD6
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 187-215 amino acids from the C-terminal region of human STARD6.
Purification Method Antigen Affinity Chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 147323
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.