Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ STARD7 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA564193
Description
Immunogen sequence: YSESSRRVLL GRLWRRLHGR PGHASALMAA LAGVFVWDEE RIQEEELQRS INEMKRLEEM SNMFQSSGVQ HHPPEPKAQT EGNEDSEGKE QRWEMVMDKK HFKL Highest antigen sequence identity to the following orthologs: Mouse - 82%, Rat - 83%.
May play a protective role in mucosal tissues by preventing exaggerated allergic responses. [UniProt].
Specifications
STARD7 | |
Polyclonal | |
Unconjugated | |
STARD7 | |
AI852671; AL022671; AW544915; gestational trophoblastic tumor protein 1; GTT1; StAR related lipid transfer domain containing 7; StARD7; StAR-related lipid transfer (START) domain containing 7; StAR-related lipid transfer domain containing 7; stAR-related lipid transfer protein 7, mitochondrial; START domain containing 7; START domain-containing protein 7 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
56910 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.8 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9NQZ5 | |
STARD7 | |
Recombinant protein corresponding to Human STARD7. Recombinant protein control fragment (Product #RP-104179). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction