Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ STARD7 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA566734
Description
Immunogen sequence: VIRPHKSFDEN GFDYLLTYSD NPQTVFPRYC VSWMVSSGMP DFLEKLHMAT LKAKNMEIKV KDYISAKPLE MSSEAKATSQ SSERKNE Highest antigen sequence identity to the following orthologs - mouse 97%, rat 95%.
May play a protective role in mucosal tissues by preventing exaggerated allergic responses. [UniProt].
Specifications
STARD7 | |
Polyclonal | |
Unconjugated | |
STARD7 | |
AI852671; AL022671; AW544915; gestational trophoblastic tumor protein 1; GTT1; StAR related lipid transfer domain containing 7; StARD7; StAR-related lipid transfer (START) domain containing 7; StAR-related lipid transfer domain containing 7; stAR-related lipid transfer protein 7, mitochondrial; START domain containing 7; START domain-containing protein 7 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
56910 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9NQZ5 | |
STARD7 | |
Recombinant Human STARD7. Recombinant protein control fragment (Product #RP-107389). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction