Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ STARD7 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5112822
Description
STARD7 Polyclonal Antibody for Western Blot, ELISA
May play a protective role in mucosal tissues by preventing exaggerated allergic responses. [UniProt].
Specifications
| STARD7 | |
| Polyclonal | |
| Unconjugated | |
| STARD7 | |
| AI852671; AL022671; AW544915; gestational trophoblastic tumor protein 1; GTT1; StAR related lipid transfer domain containing 7; StARD7; StAR-related lipid transfer (START) domain containing 7; StAR-related lipid transfer domain containing 7; stAR-related lipid transfer protein 7, mitochondrial; START domain containing 7; START domain-containing protein 7 | |
| Rabbit | |
| Protein G | |
| RUO | |
| 56910 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| Q9NQZ5 | |
| STARD7 | |
| Recombinant Human StAR-related lipid transfer protein 7, mitochondrial protein (61-307AA). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction