Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ STARD8 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA563900
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA563900 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA563900 Supplier Invitrogen™ Supplier No. PA563900
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PAQDSEQEAH SGGEPTFASS LSVEEGHSIS DTVASSSELD SSGNSMNEAE AAGPLAGLQA SMPRERRDSG V Highest antigen sequence identity to the following orthologs: Mouse - 80%, Rat - 73%.

This gene encodes a member of a subfamily of Rho GTPase activating proteins that contain a steroidogenic acute regulatory protein related lipid transfer domain. The encoded protein localizes to focal adhesions and may be involved in regulating cell morphology. This protein may also function as a tumor suppressor.
TRUSTED_SUSTAINABILITY

Specifications

Antigen STARD8
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene STARD8
Gene Accession No. Q92502
Gene Alias ARHGAP38; Deleted in liver cancer 3 protein; DLC3; DLC-3; KIAA0189; mKIAA0189; RGD1560770; StAR related lipid transfer domain containing 8; Stard8; StAR-related lipid transfer (START) domain containing 8; StAR-related lipid transfer domain containing 8; stAR-related lipid transfer protein 8; START domain containing 8; START domain-containing protein 8; STARTGAP3; START-GAP3
Gene Symbols STARD8
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human STARD8. Recombinant protein control fragment (Product #RP-100387).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9754
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.