Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Stargazin Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA14709
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA14709 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA14709 Supplier Invitrogen™ Supplier No. PA14709
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This antibody is predicted to react with mouse based on 100% sequence homology. In Western blot, PA1-4709 detects the ~36 kDa Stargazin protein.

Stargazin is a member of the transmembrane AMPAR regulatory proteins (TARP) family and is involved in glutamate receptor trafficking. It has been recently demonstrated that the interaction between stargazin and AMPA receptors is critical for the correct localization of the receptors at the synapse. Phosphorylation of the stargazin protein at Thr 321 by Protein Kinase A regulates its interaction with PSD-95 and synaptic targeting of AMPA receptors.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Stargazin
Applications Western Blot
Classification Polyclonal
Concentration 0.25 mg/mL
Conjugate Unconjugated
Formulation 0.01M HEPES with 100μg/mL BSA, 50% glycerol, 0.15M NaCl and no preservative; pH 7.5
Gene Cacng2
Gene Accession No. Q71RJ2
Gene Alias AW060990; B230105C07Rik; B930041E13Rik; CACNG2; calcium channel, voltage-dependent, gamma subunit 2; calcium voltage-gated channel auxiliary subunit gamma 2; CaV gamma 2; Ipr328; MGC123981; MGC138502; MGC138504; MRD10; Neuronal voltage-gated calcium channel gamma-2 subunit; stargazer; Stargazin; stg; TARP; TARP gamma 2; TARP gamma-2; Transmembrane AMPAR regulatory protein gamma-2; voltage-dependent calcium channel gamma-2 subunit; voltage-dependent calcium channel subunit gamma; wag; waggler
Gene Symbols Cacng2
Host Species Rabbit
Immunogen Synthetic peptide corresponding to the C-terminal region of mouse CACNG2 conjugated to KLH.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84347
Target Species Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.