Learn More
Invitrogen™ Stargazin Recombinant Rabbit Monoclonal Antibody (HL2268)

Description
Stargazin Recombinant Monoclonal Antibody for Western Blot, ICC/IF, IHC (P)
Specifications
Specifications
| Antigen | Stargazin |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Clone | HL2268 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative |
| Gene | Cacng2 |
| Gene Accession No. | O88602, Q71RJ2, Q9Y698 |
| Gene Alias | AW060990; B230105C07Rik; B930041E13Rik; CACNG2; calcium channel, voltage-dependent, gamma subunit 2; calcium voltage-gated channel auxiliary subunit gamma 2; CaV gamma 2; Ipr328; MGC123981; MGC138502; MGC138504; MRD10; Neuronal voltage-gated calcium channel gamma-2 subunit; stargazer; Stargazin; stg; TARP; TARP gamma 2; TARP gamma-2; Transmembrane AMPAR regulatory protein gamma-2; voltage-dependent calcium channel gamma-2 subunit; voltage-dependent calcium channel subunit gamma; wag; waggler |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.