Learn More
Starplex™ Scientific Histoplex™ Histology Containers
Catalog No. 1518282
Click to view available options
Capacity (Metric):
40 mL
60 mL
90 mL
120 mL
250 mL
500 mL
1 L
2 L
2.55 L
5 L
8 L
Closure Type:
Screw Cap with O-Ring
Snap Cap
Closure Color:
Clear
White
Yellow
Label:
Affixed
In case
This item is not returnable. View return policy
This item is not returnable. View return policy
Specifically engineered for the storage of histology specimens
  • O-ring polyethylene screw cap forms a seal that reduces most formalin leakage and evaporation
  • Durable polypropylene containers are translucent, flexible, resist cracking and splitting, and eliminate problems associated with formalin evaporation
  • 40mL to 1L containers have graduations for easy fill line viewing
  • 2 to 8L containers are white polypropylene with unique tamper-evident snap-on lids
Specifications
Graduated Yes
Material Polypropylene
Sterility Non-sterile
Capacity (Metric) 2.55 L
Closure Color Clear
Closure Type Snap Cap
Color Clear
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.