Learn More
Starplex™ Scientific Midstream Collection Kit
Click to view available options
Volume (Metric):
90 mL
Specifications
Specifications
Sterility | Sterile |
For Use With (Application) | Midstream Collection |
Includes | 90 mL container, magenta lined cap, funnel, castille wipe, Separate patient label and instructions enclosed in plastic bag |
No. per Case | 50/Cs. |
Volume (Metric) | 90 mL |