Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Starplex™ Scientific Starswab II™ Collection and Transport Systems: Amies Clear Medium

Catalog No. 1490567 Shop All Starplex Scientific Products
Click to view available options
Media Type:
Liquid Amies
Modified Amies Clear Gel
Shaft Material:
Plastic
Straight Wire
Swab Configuration:
Dual Applicator
Minitip flocked
Single Applicator
This item is not returnable. View return policy

Catalog No. 14-905-67

Supplier: Starplex™ Scientific SP130X

Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Protective overskirt on cap

  • Designed for collecting, transporting and maintaining the viability of aerobic, anaerobic and facultative microorganisms
  • Swab tips available in single, double or straight wire format
  • Applicators have color-coded caps with overskirt and biohazard plug-cap rings; extra-long swab shafts; and a labeled, round-bottom, spiral transport tube
  • Transport tubes contain 4mL of semi-solid media
  • Spiral media chamber within a seamless polypropylene tube protects sample and keeps it immersed in media during transport
  • Labels preprinted with lot number, expiration date and product code with space reserved for recording patient information
  • Gamma sterilized and wrapped in sterile, easy-to-peel envelope

Amies Clear Medium

  • General-purpose transport medium for recovery of a wide range of organisms, including certain anaerobes
  • Provides excellent survival characteristics for transport and gram stain compatibility

Specifications

Product Type Collection and Transport System
Media Type Modified Amies Clear Gel
Swab Configuration Single Applicator
Certifications/Compliance CE
Closure Color Magenta
For Use With (Application) Collecting, transporting and maintaining the viability of aerobic, anaerobic and facultative microorganisms
Length (English) 6 in.
Length (Metric) 15.2 cm
Packaging Bag
Quantity 50 Pk.
Shaft Material Plastic
Storage Requirements 4°C to 25°C
Tip Material Rayon
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.