Learn More
OHAUS™ Starter™ pH Electrode
Catalog No. p-7222676
Click to view available options
Junction:
Ceramic
Fiber Pin
Open
Material:
Glass
Plastic
pH Range:
0 to 14
2 to 13
The Starter™ pH Electrode features a durable shaft built to withstand daily use. Includes a temperature sensor, which powers automatic temperature compensation and ensures accurate measurements.
  • Designed for accurate and precise measurement
  • Can be used with both bench and portable meters
  • Temperature sensor features automatic temperature compensation
TRUSTED_SUSTAINABILITY
Specifications
Connector Types BNC
Junction Fiber Pin
Material Plastic
pH Range 0 to 14
Temperature (Metric) Operating 5°C to 60°C
Type pH Electrode
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.