Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
OHAUS™ Starter™ pH Meter ST2200
Shop All OHAUS Products

Click to view available options
Type:
pH Meter with Kit
Description
- Equipped with a large 3.5 in. LCD display
- Features a 99 item memory for measurements and last calibration
- Features auto-buffer recognition and auto-endpoint modes that make measurement and calibration simple
- Choose between just the ST2200 meter with electrode holder and a full kit including ST2200 meter, electrode holder, ST320 electrode, 4 pH buffer mini kits (50 mL)

Specifications
Specifications
Accuracy (pH) | ±0.01 |
Accuracy (Temperature) | ±0.5°C |
Accuracy (mV) | ±1.5 |
Data Management Details | Datalog for Measurements and Last Calibration |
Includes | ST2200 Meter, Electrode Holder, ST320 Electrode, 4 pH Buffer Mini Kits (50 mL) |
Measurement Modes | Auto-stop, Continuous |
Calibration | Linear |
Range (pH) | 0 to 14 |
Resolution (pH) | 0.01 |
Range (mV) | ±1999 |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction