Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hand2Mind Starter Science Flasks
SCI_ED

Catalog No. S02765
Click to view available options
Class Size:
1 Student

Our Starter Science Flasks are made from clear and durable plastic, feature rainbow stoppers that fit snugly, and are built to withstand years and years of experimenting.

  • Our Starter Science Flasks are made from clear and durable plastic, feature rainbow stoppers that fit snugly, and are built to withstand years and years of experimenting.

Specifications

Grade Level Elementary, Intermediate - Middle School, High School
Grade K to 12
Class Size 1 Student
Product Type Flasks
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.