Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hand2Mind Starter Science Funnels set
SCI_ED

Catalog No. S02764
Click to view available options
Class Size:
1 Student

Compatible with both solids and liquids, these funnels allow students to explore flow rates, volume, sifting, and more. Set includes 4 funnels (2 oz., 4 oz., 5 oz., and an 8 oz. powder funnel with a sifter attachment). Adjustable stand keeps funnels stable when pouring.

  • Our funnels are made from clear, durable plastic and are specially designed to demonstrate varying flow rates of liquids using different striations and shapes.

Specifications

Grade Level Elementary, Intermediate - Middle School, High School
Grade K to 12
Class Size 1 Student
Product Type Funnels
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.