Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Hand2Mind Starter Science Lab Tools Kit

Click to view available options
Grade:
K to 12
Description
- Bursting with high-quality, authentic lab tools, young scientists love this at-home set.
- Experiment Cards introduce children to scientific concepts while guiding them through experiments they can do with the tools in the kit and materials found around the house.
- Teacher-developed Activity Guide includes real-world connections to experiments, safety information, and instructions on how to use the kit components.
Specifications
Specifications
Grade Level | Elementary, Intermediate - Middle School, High School |
Grade | K to 12 |
Class Size | 1 Student |
Product Type | Starter Lab Tools Kit |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction