Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
STAT1 Antibody (246523) [Janelia Fluor™ 549], Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals FAB1490I
Description
STAT1 Monoclonal antibody specifically detects STAT1 in Human samples. It is validated for Flow Cytometry (Intracellular Staining), Immunocytochemistry, CyTOFSpecifications
STAT1 | |
Monoclonal | |
Janelia Fluor 549 | |
50mM Sodium Borate | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry (Intracellular Staining), Immunocytochemistry, CyTOF | |
246523 | |
Intracellular Staining by Flow Cytometry, Immunocytochemistry, CyTOF-ready | |
DKFZp686B04100, ISGF-3,91kD, signal transducer and activator of transcription 1, 91kDa, signal transducer and activator of transcription-1 | |
E. coli-derived recombinant human STAT1, Met1-Gln194, Accession # P42224 | |
0.1 mL | |
Apoptosis, Breast Cancer, Cancer, Myeloid derived Suppressor Cell, Signal Transduction, Transcription Factors and Regulators | |
6772 | |
Store at 4°C in the dark. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction