Learn More
STAT1 Antibody (246523), mFluor Violet 450 SE, Novus Biologicals™
Rat Monoclonal Antibody
Supplier: Novus Biologicals FAB1490MFV450
Description
STAT1 Monoclonal antibody specifically detects STAT1 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, CyTOFSpecifications
| STAT1 | |
| Monoclonal | |
| mFluor Violet 450 SE | |
| 50mM Sodium Borate | |
| Rat | |
| 0.1 mL | |
| Apoptosis, Breast Cancer, Cancer, Myeloid derived Suppressor Cell, Phospho Specific, Signal Transduction, Transcription Factors and Regulators | |
| 6772 | |
| Store at 4C in the dark. |
| Flow Cytometry, Immunocytochemistry, CyTOF | |
| 246523 | |
| Intracellular Staining by Flow Cytometry, Immunocytochemistry, CyTOF-ready | |
| DKFZp686B04100, ISGF-3,91kD, signal transducer and activator of transcription 1, 91kDa, signal transducer and activator of transcription-1 | |
| E. coli-derived recombinant human STAT1, Met1-Gln194, Accession # P42224 | |
| RUO | |
| Primary | |
| Human | |
| IgG2b |