Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STAT1 Mouse anti-Human, Monkey, Clone: 15H3, Abnova™

Catalog No. 89035566 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-035-566 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-035-566 Supplier Abnova Corporation Supplier No. MAB14168

Mouse Monoclonal Antibody

Mouse monoclonal antibody raised against human STAT1.

  • High specificity
  • High reactivity

Specifications

Antigen STAT1
Applications Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 15H3
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against human STAT1.
Dilution Immunocytochemistry (1:20) Immunofluorescence (1:20) Immunoprecipitation (2 ug) Western Blot (1:1000) The optimal working dilution should be determined by the end user.
Formulation In PBS (30% glycerol, 1 mg/mL BSA, 0.05% sodium azide).
Gene Accession No. P42224
Gene Alias DKFZp686B04100/ISGF-3/STAT91
Gene Symbols STAT1
Host Species Mouse
Immunogen Protein expressed in 293T cell transfected with human STAT1 expression vector.
Purification Method Protein A affinity purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6772
Target Species Human, Monkey
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.