Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
STAT1, p Ser727 Antibody (PSM1), Alexa Fluor™ 532, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB500515AF532
Description
STAT1 Monoclonal specifically detects STAT1 in Human samples. It is validated for Western Blot.Specifications
STAT1 | |
Monoclonal | |
Alexa Fluor 532 | |
50 mM sodium borate with 0.05% sodium azide | |
STAT1 | |
STAT1 peptide sequence 721-733 (Ser727 phosphorylated). | |
Primary | |
The antibody PSM1 recognizes transcriptional factor STAT1 (91 kDa) activated by phosphorylation at Ser727. | |
Store at 4°C in the dark. |
Western Blot | |
PSM1 | |
Western Blot | |
DKFZp686B04100, ISGF-3,91kD, signal transducer and activator of transcription 1, 91kDa, signal transducer and activator of transcription-1 | |
Mouse | |
0.1 mL | |
6772 | |
Human | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction