Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Stat1 (pY701) Mouse anti-Human, Mouse, Rat, RB780, Clone: 4a, BD Phosflow™

Catalog No. BDB569144 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
25 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB569144 100 Tests
BDB569145 25 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB569144 Supplier BD Biosciences Supplier No. 569144
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Stat1 (pY701) Monoclonal antibody specifically detects Stat1 (pY701) in Human|Mouse|Rat samples. It is validated for Intracellular staining (flow cytometry)
TRUSTED_SUSTAINABILITY

Specifications

Antigen Stat1 (pY701)
Applications Flow Cytometry, Control
Classification Monoclonal
Clone 4a
Conjugate RB780
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias Signal Transducer and Activator of Transcription-1; STAT91; ISGF-3; CANDF7
Host Species Mouse
Immunogen Phosphorylated Human Stat1 Peptide
Purification Method Affinity Chromatography
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human, Mouse, Rat
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions and unreacted dye was removed.
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.