Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STAT1, Rabbit anti-Human, Clone: C6, Abnova™

Catalog No. 89335939 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-335-939 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-335-939 Supplier Abnova Corporation Supplier No. MAB19105
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Specifications

Antigen STAT1
Applications Flow Cytometry, Western Blot
Classification Monoclonal
Clone C6
Conjugate Unconjugated
Dilution Flow Cytometry (1 ug/mL-0.001 ug/mL ) Western Blot The optimal working dilution should be determined by the end user.
Formulation In PBS (50% glycerol, 0.1% BSA, 0.02% sodium azide).
Gene signal transducer and activator of transcription 1, 91kDa
Gene Alias DKFZp686B04100/ISGF-3/STAT91
Gene Symbols STAT1
Host Species Rabbit
Immunogen A synthetic phosphopeptide corresponding to residues surrounding Ser727 of human phospho STAT1.
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6772
Target Species Human
Content And Storage Store at -20°C. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.