Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
STAT1, Rabbit anti-Human, Clone: C6, Abnova™
Rabbit Monoclonal Antibody
Supplier: Abnova Corporation MAB19106
Specifications
STAT1 | |
Monoclonal | |
Unconjugated | |
In PBS (50% glycerol, 0.1% BSA, 0.02% sodium azide). | |
DKFZp686B04100/ISGF-3/STAT91 | |
Rabbit | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG1 κ |
Flow Cytometry, Western Blot | |
C6 | |
Flow Cytometry (1 ug/mL-0.001 ug/mL ) Western Blot The optimal working dilution should be determined by the end user. | |
signal transducer and activator of transcription 1, 91kDa | |
STAT1 | |
A synthetic phosphopeptide corresponding to residues surrounding Ser727 of human phospho STAT1. | |
RUO | |
6772 | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction