Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STAT1, Rabbit anti-Human, SureLight 488, Clone: C6, Abnova™

Catalog No. 89335947 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 Reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-335-947 100 Reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-335-947 Supplier Abnova Corporation Supplier No. MAB19113
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Specifications

Antigen STAT1
Applications Flow Cytometry
Classification Monoclonal
Clone C6
Conjugate SureLight 488
Dilution Flow Cytometry (5 uL/106 cells or 5 u per 100 uL) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (0.2% BSA, 0.09% sodium azide).
Gene signal transducer and activator of transcription 1, 91kDa
Gene Alias DKFZp686B04100/ISGF-3/STAT91
Gene Symbols STAT1
Host Species Rabbit
Immunogen A synthetic phosphopeptide corresponding to residues surrounding Ser727 of human phospho STAT1.
Quantity 100 Reactions
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6772
Target Species Human
Content And Storage Store at 4°C.
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.