Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
STAT2 Antibody (STAT2/2650) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP308587
Description
STAT2 Monoclonal specifically detects STAT2 in Human samples. It is validated for Immunohistochemistry-Paraffin.Specifications
STAT2 | |
Monoclonal | |
1 mg/mL | |
Immunohistochemistry-Paraffin : 1-2 ug/mL | |
interferon alpha induced transcriptional activator, ISGF-3, MGC59816, p113, signal transducer and activator of transcription 2, signal transducer and activator of transcription 2, 113kD, signal transducer and activator of transcription 2, 113kDa, STAT113 | |
Recombinant full-length human STAT2 protein | |
100 μg | |
Cancer, Phospho Specific, Signal Transduction, Transcription Factors and Regulators | |
6773 | |
Store at -20 to -80C. Avoid freeze-thaw cycles. | |
IgG1 κ |
Immunohistochemistry (Paraffin) | |
STAT2/2650 | |
Unconjugated | |
10mM PBS | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction