Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
STAT2, p Tyr689 Antibody (1021D), Allophycocyanin/Cy7, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals FAB2890APCCY7
Description
STAT2 Monoclonal antibody specifically detects STAT2 in Human samples. It is validated for Flow CytometrySpecifications
STAT2 | |
Monoclonal | |
APC-Cyanine7 | |
PBS | |
Rabbit | |
Protein A or G purified from cell culture supernatant | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry | |
1021D | |
Flow Cytometry | |
interferon alpha induced transcriptional activator, ISGF-3, MGC59816, p113, signal transducer and activator of transcription 2, signal transducer and activator of transcription 2, 113kD, signal transducer and activator of transcription 2, 113kDa, STAT113 | |
Phosphopeptide containing the human STAT2 Y690 site | |
0.1 mL | |
Cancer, Phospho Specific, Signal Transduction, Transcription Factors and Regulators | |
6773 | |
Store at 4C in the dark. Do not freeze. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction