Learn More
Invitrogen™ STAT2 Recombinant Rabbit Monoclonal Antibody (4A5P8)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA542828
Description
Immunogen sequence: KLPFWTWLDK ILELVHDHLK DLWNDGRIMG FVSRSQERRL LKKTMSGTFL LRFSESSEGG ITCSWVEHQD DDKVLIYSVQ PYTKEVLQSL PLTEIIRHYQ LLTEENIPEN PLRFLYPRIP RDEAFGCYYQ EKVNLQERRK YLKHRLIVVS NRQVDEL; Positive test controls include: A-549, U-937, Mouse brain, Rat testis. The target is usually found in the following locations: Cytoplasm, Nucleus.
STAT2 is a member of the STAT protein family. In response to cytokines and growth factors, STAT family members are phosphorylated by the receptor associated kinases, and then form homo- or heterodimers that translocate to the cell nucleus where they act as transcription activators. In response to interferon (IFN), this protein forms a complex with STAT1 and IFN regulatory factor family protein p48 (ISGF3G), in which this protein acts as a transactivator, but lacks the ability to bind DNA directly. Transcription adaptor P300/CBP (EP300/CREBBP) has been shown to interact specifically with this protein, which is thought to be involved in the process of blocking IFN-alpha response by adenovirus.
Specifications
STAT2 | |
Recombinant Monoclonal | |
1 mg/mL | |
PBS with 0.05% BSA, 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
P52630 | |
Stat2 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 550-706 of human STAT2 (NP_0054101). | |
100 μL | |
Primary | |
Human, Rat | |
Antibody | |
IgG |
ELISA, Western Blot | |
4A5P8 | |
Unconjugated | |
Stat2 | |
1110034C02Rik; 1600010G07Rik; acute phase response factor; acute-phase response factor; ADMIO; ADMIO1; Aprf; AW109958; AW496480; DNA-binding protein APRF; EGK_03803; HIES; IMD44; interferon alpha induced transcriptional activator; ISGF-3; MGC59816; p113; signal transducer and activator of transcription 2; signal transducer and activator of transcription 2, 113kDa; signal transducer and activator of transcription 3; signal transducer and activator of transcription 3 (acute-phase response factor); STAT; STAT 2; STAT113; STAT2; STAT-2; Stat3 | |
Rabbit | |
Affinity chromatography | |
RUO | |
288774, 6773 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.