Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
STAT3 Antibody (232209), Allophycocyanin, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB1799A
Description
STAT3 Monoclonal antibody specifically detects STAT3 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunoprecipitation, Flow Cytometry (Intracellular Staining), Immunocytochemistry, Gene Knock-OutSpecifications
STAT3 | |
Monoclonal | |
APC | |
PBS | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot, Immunoprecipitation, Flow Cytometry (Intracellular Staining), Immunocytochemistry, Gene Knock-Out | |
232209 | |
Western Blot, Immunoprecipitation, Intracellular Staining by Flow Cytometry, Immunocytochemistry, Knockout Validated | |
Acute-phase response factor, APRFMGC16063, DNA-binding protein APRF, FLJ20882, HIES, signal transducer and activator of transcription 3, signal transducer and activator of transcription 3 (acute-phase responsefactor) | |
E. coli-derived recombinant human STAT3, Met1-Asn175, Accession # P40763 | |
0.1 mL | |
Cellular Markers, Myeloid derived Suppressor Cell, Signal Transduction, Transcription Factors and Regulators | |
6774 | |
Store at 4°C in the dark. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction