Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
STAT3 Antibody (OTI21E7), DyLight 680, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP274378FR
Description
STAT3 Monoclonal specifically detects STAT3 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Knockout Validated.Specifications
STAT3 | |
Monoclonal | |
DyLight 680 | |
PBS with 0.05% Sodium Azide | |
STAT3 | |
Recombinant protein expressed in E.coli corresponding to amino acids 154-175 of human STAT3. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Western Blot, Immunohistochemistry, Immunoassay | |
OTI21E7 | |
Western Blot, Immunohistochemistry, Knockout Validated | |
Acute-phase response factor, APRFMGC16063, DNA-binding protein APRF, FLJ20882, HIES, signal transducer and activator of transcription 3, signal transducer and activator of transcription 3 (acute-phase responsefactor) | |
Mouse | |
Affinity Purified | |
Cellular Markers, Myeloid derived Suppressor Cell, Phospho Specific, Signal Transduction, Transcription Factors and Regulators | |
6774.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction