Learn More
Invitrogen™ STAT3 Monoclonal Antibody (232209), FITC

Description
STAT3 Monoclonal Antibody for Flow
Specifications
Specifications
| Antigen | STAT3 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | 232209 |
| Conjugate | FITC |
| Formulation | Buffered saline solution with 0.5% BSA and 0.09% sodium azide |
| Gene | STAT3 |
| Gene Accession No. | P40763, P42227, P52631 |
| Gene Alias | 1110034C02Rik; acute phase response factor; acute-phase response factor; ADMIO; ADMIO1; APRF; AW109958; DNA-binding protein APRF; FLJ20882; HIES; MGC16063; signal transducer and activator of transcription 3; signal transducer and activator of transcription 3 (acute-phase response factor); signal transducer; activator of transcription; acute-phase response factor; signal transduction and activation of transcription 3; STAT; STAT3; STAT-3; stat3 protein; STAT3b1; STAT3b2; transcription factor; Unknown (protein for MGC:128731); wu:fc15d02; wu:fl59g06; z-Stat3 |
| Gene Symbols | STAT3 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.