Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
STAT3, p Tyr705 Antibody (1004G), DyLight 488, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals FAB46071K
Description
STAT3 Monoclonal antibody specifically detects STAT3 in Human samples. It is validated for Western Blot, Flow Cytometry (Intracellular Staining)Specifications
| STAT3 | |
| Monoclonal | |
| DyLight 488 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Flow Cytometry (Intracellular Staining) | |
| 1004G | |
| Western Blot, Intracellular Staining by Flow Cytometry | |
| Acute-phase response factor, APRFMGC16063, DNA-binding protein APRF, FLJ20882, HIES, signal transducer and activator of transcription 3, signal transducer and activator of transcription 3 (acute-phase responsefactor) | |
| Phosphopeptide containing the human STAT3 Y705 site, Accession # P40763 | |
| 0.1 mL | |
| Cellular Markers, Myeloid derived Suppressor Cell, Phospho Specific, Signal Transduction, Transcription Factors and Regulators | |
| 6774 | |
| Store at 4°C in the dark. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction