Learn More
Invitrogen™ STAT3 Polyclonal Antibody

Description
Immunogen sequence: EKPMEIARIV ARCLWEESRL LQTAATAAQQ GGQANHPTAA VVTEKQQMLE QHLQDVRKRV QDLEQKMKVV ENLQDDFDFN YKTLKSQGDM QDLNGNNQSV TRQKMQQLEQ MLTALDQMRR SIVSELAGLL SAMEYVQK Highest antigen sequence indentity to the following orthologs: Rat - 100%, Mouse - 100%.
Specifications
Specifications
| Antigen | STAT3 |
| Applications | ChIP Assay, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 0.10 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | STAT3 |
| Gene Accession No. | P40763 |
| Gene Alias | 1110034C02Rik; acute phase response factor; acute-phase response factor; ADMIO; ADMIO1; APRF; AW109958; DNA-binding protein APRF; FLJ20882; HIES; MGC16063; signal transducer and activator of transcription 3; signal transducer and activator of transcription 3 (acute-phase response factor); signal transducer; activator of transcription; acute-phase response factor; signal transduction and activation of transcription 3; STAT; STAT3; STAT-3; stat3 protein; STAT3b1; STAT3b2; transcription factor; Unknown (protein for MGC:128731); wu:fc15d02; wu:fl59g06; z-Stat3 |
| Gene Symbols | STAT3 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.