Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Stat3 (pY705) Mouse, Alexa Fluor 647, Clone: 4/P-STAT3, BD

Catalog No. BDB562071 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
250 Tests
50 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB562071 250 Tests
BDB557815 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB562071 Supplier BD Biosciences Supplier No. 562071
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

STAT (Signal transducer and activator of transcription) of Y705 in Stat3 occurs in response to growth factors and cytokines, and is essential for normal transcription activity.

The 4/P-STAT3 monoclonal antibody recognizes the phosphorylated Y705 of Stat3.

TRUSTED_SUSTAINABILITY

Specifications

Antigen Stat3 (pY705)
Applications Flow Cytometry
Classification Monoclonal
Clone 4/P-STAT3
Conjugate Alexa Fluor 647
Description Acute-phase response factor, APRF
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Phosphorylated Human Stat3 Peptide
Purification Method Affinity Purified
Quantity 250 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human, Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2a κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.