Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STAT3 Rabbit anti-Human, Mouse, PE, Clone: B12, Abnova™

Catalog No. 89335963 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 Reactions
100 Reactions
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-335-963 100 Reactions
89-335-964 10 Reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-335-963 Supplier Abnova Corporation Supplier No. MAB19129
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody.

STAT3 Monoclonal antibody specifically detects STAT3 in Human, Mouse samples. It is validated for Flow Cyt.

Specifications

Antigen STAT3
Applications Flow Cytometry
Classification Monoclonal
Clone B12
Conjugate PE
Description signal transducer and activator of transcription 3 (acute-phase response factor)
Gene STAT3
Gene Alias APRF, FLJ20882, HIES, MGC16063
Host Species Rabbit
Immunogen A synthetic phosphopeptide corresponding to residues surrounding Y705 of human STAT3
Quantity 100 Reactions
Primary or Secondary Primary
Gene ID (Entrez) 6774
Target Species Human, Mouse
Content And Storage Store at 4°C
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.