Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Stat3b, Mouse, Unlabeled, Clone: 516 G10H9,

Catalog No. MABC1177MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
MABC1177MI 100 μg Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABC1177MI Supplier MilliporeSigma™ Supplier No. MABC1177
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Signal transducer and activator of transcription 3 (UniProt P40763; also known as Acute-phase response factor, DNA-binding protein APRF) is encoded by the STAT3 (also known as ADMIO, APRF, HIES) gene (Gene ID 6774) in human. STAT proteins (1, 2, 3, 4, 5A, 5B, 6) constitute a family of proteins that mediate cytokine and growth factor receptors-initiated signaling events. Stat3 is activated upon phosphorylation on Y705 by Jak or receptor tyrosine kinases, leading to Stat3 homodimerization, nuclear translocation, and target genes transcription. Constitutively tyrosine-phosphorylated Stat3 is implicated in the initiation and progression of many types of cancers, while Stat3 gene knockout in mice is lethal at the early embryonic stage. Two major alternatively spliced Stat3 isoforms exist, the C-terminal end 55-amino acid sequence (a.a. 716-770) in the canonical alpha isoform (UniProt P40763-1) is truncated and replaced with a 7-amino acid sequence in the 722-amino acid beta isoform (UniProt P40763-3). Tyrosine-phosphorylated Stat3β homodimers are more stable and less susceptible to dephosphorylation, bind DNA more avidly and exhibit markedly prolonged nuclear retention than pStat3α homodimers.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Stat3b
Applications Western Blot
Classification Monoclonal
Clone 516 G10H9
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG1κ antibody in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% Sodium Azide.
Gene STAT3, ADMIO, APRF, HIES
Gene Accession No. P40763-3
Host Species Mouse
Immunogen Linear peptide containing human Stat3b C-terminal end 7 a.a. (CT7).
Purification Method Protein G Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Apoptosis & Cancer
Primary or Secondary Primary
Gene ID (Entrez) NP_998827
Test Specificity Clone 516 G10H9 detects the C-terminal end 7-a.a. (CT7) sequence present in human/mouse spliced isoform Stat3B (UniProt P40763-3/P42227-2), but not in human/mouse spliced isoform Stat3A (UniProt P40763-1/P42227-1) or human/mouse isoform Del-701 (UniProt P40763-2/P42227-3).
Target Species Human
Content And Storage Stable for 1 year at 2°-8°C from date of receipt.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.