Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
STAT4 Antibody (OTI2F3), FITC, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP274389F
Description
STAT4 Monoclonal specifically detects STAT4 in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
STAT4 | |
Monoclonal | |
FITC | |
PBS with 0.05% Sodium Azide | |
STAT4 | |
Full length recombinant protein of human STAT4 (NP_003142) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
OTI2F3 | |
Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
signal transducer and activator of transcription 4, SLEB11 | |
Mouse | |
Affinity Purified | |
Cancer, Phospho Specific, Regulatory Immunology, Signal Transduction | |
6775.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction