Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STAT4 Monoclonal Antibody (OTI4F9), TrueMAB™, OriGene
SDP

Catalog No. 502541167
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-1167 30 μL
50-167-4000 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-254-1167 Supplier OriGene Supplier No. TA503022S
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

STAT4 Monoclonal antibody specifically detects STAT4 in Canine, Human, Rat samples. It is validated for Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen STAT4
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI4F9
Concentration 0.28 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene AP2M1
Gene Accession No. P84092, Q96CW1
Gene Alias CIS4; HSPC060; OTTHUMP00000163553; Signal transducer and activator of transcription 4; SLEB11; SOCS4; SOCS6; SSI4; STAI4; STAT; Stat4; STATI4
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 97-383 of human AP2M1 produced in E.coli.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 116563, 1173, 607139
Target Species Canine, Human, Rat
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.