Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STAT4 Mouse anti-Human, Biotin, Clone: OTI2G5, liquid, TrueMAB™
SDP

Catalog No. 501677714
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-167-7714 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-167-7714 Supplier OriGene Supplier No. TA700193
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The STAT 4 protein is a transcription factor activated by hormone and cytokines. STAT 4 is most similar to STAT 1 (52%) and STAT 3 (47%). Functionally, STAT 4 is similar to other STAT family members in that it can be tyrosine phosphorylated by JAK 1 or JAK 2. STAT 4 forms homodimers and heterodimers with related STAT family members. Tyrosine phosphorylated STAT 4 can bind the interferon g activated site (GAS), and serine phosphorylation is also required for maximal transcriptional activity. STAT 4 expression is restricted to the thymus, spleen, and testis.

Specifications

Antigen STAT4
Applications ELISA
Classification Monoclonal
Clone OTI2G5
Concentration 0.5 mg/mL
Conjugate Biotin
Formulation PBS with 10mg/ml BSA, 50% glycerol and 0.05% sodium azide
Gene STAT4
Gene Accession No. Q14765
Gene Alias SLEB11
Gene Symbols STAT4
Host Species Mouse
Immunogen Full length human recombit protein of human STAT4 produced in HEK293T cell.
Purification Method Affinity Chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6775
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.