Learn More
Stat4 (pY693) Mouse, PE, Clone: 38/P-STAT4, BD
Mouse Monoclonal Antibody
$317.50 - $1430.00
Specifications
Antigen | Stat4 (pY693) |
---|---|
Clone | 38/P-STAT4 |
Applications | Flow Cytometry |
Classification | Monoclonal |
Conjugate | PE |
Description
The Stat proteins function both as cytoplasmic signal transducers and as activators of transcription. Seven mammalian Stat proteins have been identified: Stat1-4, Stat5a, 5b, and Stat6. Stat4 has been shown to play an important role in development of T helper cells, specifically the Th1 subset. Stat4 is activated by IL-12 and by type 1 interferons. Knockout mice supported the role that Stat4 plays in IL-12 signaling because lymphocytes from Stat 4-/- mice could neither differentiate into Th1 cells nor produce IFN-γ in response to treatment with IL-12. IFN-γ plays an important role in host defense. A key component in the activation of Stat4 is the phosphorylation on tyrosine and serine residues in response to IL-12 stimulation. IL-12 stimulation leads to the phosphorylation of Stat4 on tyrosine 693 and serine 721. Transcriptional activity of Stat4 has been shown to be significantly reduced when residues Y693 and S721 are mutated.
Specifications
Stat4 (pY693) | |
Flow Cytometry | |
PE | |
RUO | |
Aqueous buffered solution containing BSA, protein stabilizer, and ≤0.09% sodium azide. | |
IgG2b κ | |
Affinity Purified | |
Signal transducer and activator of transcription 4; SLEB11 |
38/P-STAT4 | |
Monoclonal | |
Mouse | |
Human, Mouse | |
Phosphorylated Human Stat4 (pY693) | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.