Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STAT4 Rabbit anti-Human, Mouse, APC, Clone: F6, Abnova™

Catalog No. 89335968 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 Reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-335-968 10 Reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-335-968 Supplier Abnova Corporation Supplier No. MAB19134
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody.

STAT4 Monoclonal antibody specifically detects STAT4 in Human, Mouse samples. It is validated for Flow Cyt.

Specifications

Antigen STAT4
Applications Flow Cytometry
Classification Monoclonal
Clone F6
Conjugate APC
Description signal transducer and activator of transcription 4
Gene STAT4
Gene Alias SLEB11
Host Species Rabbit
Immunogen A synthetic phosphopeptide corresponding to residues surrounding Y693 of human STAT4
Quantity 10 Reactions
Primary or Secondary Primary
Gene ID (Entrez) 6775
Target Species Human, Mouse
Content And Storage Store at 4°C
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.