Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STAT5 Polyclonal Antibody, Abbomax

Catalog No. 502203825
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-220-3825 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-220-3825 Supplier Abbomax Inc Supplier No. 5008364
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive control: Mouse liver Cellular location: Nuclear.

STAT5 Polyclonal antibody specifically detects STAT5 in Human, Mouse samples. It is validated for Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen STAT5
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with proprietary stabilizer and 0.01% sodium azide
Gene STAT5B
Gene Accession No. P42229, P42230, P42232, P51692
Gene Alias AA959963; Mammary gland factor; mammary gland factor STAT5A; mammary gland factor STAT5B; mammary gland factor; transcription factor; MGF; Mpf; signal transducer and activator of transcription 5; Signal transducer and activator of transcription 5a; Signal transducer and activator of transcription 5B; STA5B; STAT; STAT5; Stat5a; Stat5b; transcription factor STAT5B
Host Species Rabbit
Immunogen A synthetic peptide surrounding to the C-terminus of STAT5 protein
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20850, 20851, 6776, 6777
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.