Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
STAT5A Antibody (251610), Alexa Fluor™ 700, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB21741N
Description
STAT5A Monoclonal antibody specifically detects STAT5A in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, CyTOFSpecifications
STAT5A | |
Monoclonal | |
Alexa Fluor 700 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry, Immunocytochemistry, CyTOF | |
251610 | |
Intracellular Staining by Flow Cytometry, Immunocytochemistry, CyTOF-ready | |
MGF, signal transducer and activator of transcription 5A, STAT5 | |
Human STAT5a synthetic peptide, SLDSRLSPPAGLFTSARGSLS, Accession # NP_003143 | |
0.1 mL | |
Cancer, Phospho Specific, Signal Transduction, Transcription Factors and Regulators | |
6776 | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction