Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
STAT5A, Asym-dimethyl Arg31 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP244185
Description
Specifically detects STAT5a (Asym-dimethyl Arg31) in Human samples, and it is validated for ELISA, Western Blotting
STAT5A Polyclonal specifically detects STAT5A in Human samples. It is validated for Western Blot, ELISA.Specifications
STAT5a (Asym-dimethyl Arg31) | |
Polyclonal | |
Unconjugated | |
0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 30% Glycerol with 0.05% Sodium Azide | |
MGF, signal transducer and activator of transcription 5A, STAT5 | |
Rabbit | |
90.65 kDa | |
0.1 mg | |
Cancer, Phospho Specific, Signal Transduction, Transcription Factors and Regulators | |
6776 | |
Store at -20°C short term. Aliquot and store at -80°C long term. Avoid freeze-thaw cycles. |
Western Blot, ELISA | |
0.75 mg/mL | |
Western Blot 1:500, ELISA 1:5000 | |
P42229 | |
STAT5A | |
STAT5A [Asym-dimethyl Arg31] Antibody was prepared from whole rabbit serum produced by repeated immunizations with a synthetic monomethylated peptide surrounding Arginine 31 of human STAT. (Uniprot: P42229) | |
Affinity Purified | |
RUO | |
Primary | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction