Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
STAT5a/b Antibody (SR2282), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP32169250UL
Description
STAT5a/b Monoclonal antibody specifically detects STAT5a/b in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, ImmunoprecipitationSpecifications
STAT5a/b | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunoprecipitation | |
SR2282 | |
Western Blot 1:1000-1:2000, Flow Cytometry 1:50, Immunohistochemistry 1:100-1:500, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunoprecipitation 1:50 | |
MGF, STAT5, STAT5A signal transducer and activator of transcription 5A | |
A synthesized peptide derived from human STAT5a/b (Uniprot #: P42229, P51692 ) | |
50 μg | |
Cancer, Phospho Specific, Signal Transduction, Transcription Factors and Regulators | |
6776 | |
Store at -20° C. Avoid freeze/thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction