Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
STAT5A, Mouse anti-Human, Clone: 9F7, Abnova™
Mouse Monoclonal Antibody
Supplier: Abnova Corporation MAB16377
Specifications
STAT5A | |
Monoclonal | |
Unconjugated | |
In PBS, pH 7.3 (50% glycerol, 1% BSA, 0.02% sodium azide). | |
MGF/STAT5 | |
Mouse | |
50 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
9F7 | |
Flow Cytometry (1:100) Immunofluorescence (1:100) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (10 ug/mL) Western Blot (1:500-1:2000) The optimal working dilution should be determined by the end user. | |
signal transducer and activator of transcription 5A | |
STAT5A | |
Recombinant protein corresponding to full length human STAT5A from HEK293T cell. | |
RUO | |
6776 | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction