Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
STAT5A Rabbit anti-Human, Mouse, APC, Clone: G11, Abnova™
Rabbit Monoclonal Antibody.
$449.50 - $688.00
Specifications
Antigen | STAT5A |
---|---|
Clone | G11 |
Applications | Flow Cytometry |
Classification | Monoclonal |
Conjugate | APC |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
89-335-978
|
Abnova Corporation
MAB19144 |
10 Reactions |
Each for $449.50
|
|
|||||
89-335-977
|
Abnova Corporation
MAB19143 |
100 Reactions |
Each for $688.00
|
|
|||||
Description
STAT5A Monoclonal antibody specifically detects STAT5A in Human, Mouse samples. It is validated for Flow Cyt.Specifications
STAT5A | |
Flow Cytometry | |
APC | |
Rabbit | |
MGF, STAT5 | |
A synthetic phosphopeptide corresponding to residues surrounding Y694 of human STAT5 | |
Primary | |
signal transducer and activator of transcription 5A |
G11 | |
Monoclonal | |
Liquid | |
Human, Mouse | |
6776, 6777 | |
IgG1 κ | |
Store at 4°C | |
STAT5A |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title