Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
STAT5A Rabbit anti-Human, Mouse, Unconjugated, Clone: G11, Abnova™
Rabbit Monoclonal Antibody.
Supplier: Abnova Corporation MAB19141
Description
STAT5A Monoclonal antibody specifically detects STAT5A in Human, Mouse samples. It is validated for WB, Flow Cyt.Specifications
STAT5A | |
Monoclonal | |
Unconjugated | |
STAT5A | |
Rabbit | |
200 μL | |
6776, 6777 | |
Store at -20°C | |
IgG1 κ |
Western Blot, Flow Cytometry | |
G11 | |
signal transducer and activator of transcription 5A | |
MGF, STAT5 | |
A synthetic phosphopeptide corresponding to residues surrounding Y694 of human STAT5 | |
Primary | |
Human, Mouse | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction