Learn More
STAT5b Antibody (389215), mFluor Violet 500 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB1584MFV500
Description
STAT5b Monoclonal antibody specifically detects STAT5b in Human samples. It is validated for Western Blot, Western Blot, Immunoprecipitation, Flow Cytometry, CyTOF, KnockDownSpecifications
| STAT5b | |
| Monoclonal | |
| mFluor Violet 500 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| 0.1 mL | |
| Phospho Specific, Signal Transduction | |
| 6777 | |
| Store at 4C in the dark. |
| Western Blot, Immunoprecipitation, Flow Cytometry, CyTOF | |
| 389215 | |
| Western Blot, Simple Western, Immunoprecipitation, Intracellular Staining by Flow Cytometry, CyTOF-ready, Knockout Validated | |
| signal transducer and activator of transcription 5B, STAT5, transcription factor STAT5B | |
| Human STAT5b synthetic peptide, MDSQWIPHAQS, Accession # NP_036580 | |
| RUO | |
| Primary | |
| Human | |
| IgG2b |