Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
STAT5b Antibody (389215) [PerCP], Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB1584C
Description
STAT5b Monoclonal antibody specifically detects STAT5b in Human samples. It is validated for Western Blot, Simple Western, Immunoprecipitation, Flow Cytometry (Intracellular Staining), CyTOF, Gene Knock-OutSpecifications
STAT5b | |
Monoclonal | |
PerCP | |
PBS | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, Simple Western, Immunoprecipitation, Flow Cytometry (Intracellular Staining), CyTOF, Gene Knock-Out | |
389215 | |
Western Blot, Simple Western, Immunoprecipitation, Intracellular Staining by Flow Cytometry, CyTOF-ready, Knockout Validated | |
signal transducer and activator of transcription 5B, STAT5, transcription factor STAT5B | |
Human STAT5b synthetic peptide, MDSQWIPHAQS, Accession # NP_036580 | |
0.1 mL | |
Signal Transduction | |
6777 | |
Store at 4°C in the dark. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction