Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
STAT5b Antibody (STAT5B/2611), FITC, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP279907F
Description
STAT5b Monoclonal specifically detects STAT5b in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
STAT5b | |
Monoclonal | |
FITC | |
PBS with 0.05% Sodium Azide | |
STAT5B | |
Recombinant full-length human STAT5b protein (Uniprot: P51692) | |
0.1 mL | |
Phospho Specific, Signal Transduction | |
Human | |
IgG1 κ |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
STAT5B/2611 | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
signal transducer and activator of transcription 5B, STAT5, transcription factor STAT5B | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction